Gene Transcripts

Gene Symbol
KRTCAP2
Entrez Gene ID
200185
Gene Name
keratinocyte associated protein 2
Gene Aliases
KCP2
Chromosome Location
Chr. 1: 155169408 - 155173328 on Build GRCh38
UniGene ID
Hs.516671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173852.3 NP_776251.1 2, 3 270
GenBank mRNA AK311907.1 - 2, 3 244
AY157577.1 - 1, 2 155
BC029806.1 - 2, 3 257 5189880
BC048205.1 - 2, 3 196
BC057233.1 - 2, 3 279 6463372
BX647396.1 - 2, 3 199

View Full Product Details