Gene Transcripts

Gene Symbol
COMMD6
Entrez Gene ID
170622
Gene Name
COMM domain containing 6
Gene Aliases
Acrg
Chromosome Location
Chr. 13: 75525214 - 75549439 on Build GRCh38
UniGene ID
Hs.508266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287392.1 NP_001274321.1 4 551
NM_001287393.1 NP_001274322.1 4 536
NM_001287394.1 NP_001274323.1 3 367
NM_203495.3 NP_987091.1 4 371
NM_203497.3 NP_987093.1 5 410
XM_011534948.2 XP_011533250.1 5 464
GenBank mRNA AF169971.1 - 4 338
AK000009.1 - 4 340
BC110837.1 - 4 308
BC117391.1 - 4 348 40125942
BC143912.1 - 4 348 9052430
BC171758.1 - 4 348 9054411
BM476717.1 - 304
BX538116.1 - 4 494
CD175553.1 - 4 371
DB487154.2 - 4 354
DB509325.2 - 3 356
HY028175.1 - 5 383
HY105279.1 - 4 372

View Full Product Details