Gene Transcripts

Gene Symbol
CATIP
Entrez Gene ID
375307
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C2orf62
Chromosome Location
Chr. 2: 218356757 - 218368393 on Build GRCh38
UniGene ID
Hs.645453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320865.1 NP_001307794.1 5 467
NM_198559.1 NP_940961.1 5 434
XM_005246539.4 XP_005246596.1 4 431
XM_005246541.4 XP_005246598.1 3 230
XM_011511148.2 XP_011509450.1 5 566
XM_011511149.2 XP_011509451.1 3 229
XM_011511150.2 XP_011509452.1 3 195
XM_017004053.1 XP_016859542.1 3 602
GenBank mRNA BC052750.1 - 5 434

View Full Product Details