Gene Transcripts

Gene Symbol
UBE3C
Entrez Gene ID
9690
Gene Name
ubiquitin protein ligase E3C
Gene Aliases
HECTH2
Chromosome Location
Chr. 7: 157138952 - 157269372 on Build GRCh38
UniGene ID
Hs.118351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014671.2 NP_055486.2 1, 2 389
XM_005249564.4 XP_005249621.1 1, 2 398
XM_017012818.1 XP_016868307.1 2, 3 661
GenBank mRNA AK295138.1 - 1, 2 69
BC014029.2 - 2, 3 149 4541634
D13635.1 - 1, 2 380
DB454915.1 - 1, 2 389

View Full Product Details