Gene Transcripts

Gene Symbol
OR8K1
Entrez Gene ID
390157
Gene Name
olfactory receptor family 8 subfamily K member 1
Gene Aliases
OR11-182, OR8N1P
Chromosome Location
Chr. 11: 56346039 - 56346998 on Build GRCh38
UniGene ID
Hs.553748
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002907.1 NP_001002907.1 1 411
GenBank mRNA BC146844.1 - 1 505

View Full Product Details