Gene Transcripts

Gene Symbol
FBXW10
Entrez Gene ID
10517
Gene Name
F-box and WD repeat domain containing 10
Gene Aliases
Fbw10, HREP, SM25H2, SM2SH2
Chromosome Location
Chr. 17: 18744013 - 18779349 on Build GRCh38
UniGene ID
Hs.592128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267585.1 NP_001254514.1 14 2605
NM_001267586.1 NP_001254515.1 13 2446
XM_011523625.1 XP_011521927.1 14 2884
GenBank mRNA AL136822.1 - 13 2384
AY729024.1 - 14 2373
BC028364.2 - 12 2619 4827344
BI828401.1 - 4 484

Gene Symbol
CDRT1
Entrez Gene ID
374286
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
C170RF1, C17ORF1, C17ORF1A, FBXW10B, FBXW10P1, HREP, SM25H2
Chromosome Location
Chr. 17: 15565482 - 15619704 on Build GRCh38
UniGene ID
Hs.548021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282540.1 NP_001269469.1 13 2373

View Full Product Details