Gene Transcripts

Gene Symbol
PLEKHA7
Entrez Gene ID
144100
Gene Name
pleckstrin homology domain containing A7
Gene Aliases
Chromosome Location
Chr. 11: 16787153 - 17014423 on Build GRCh38
UniGene ID
Hs.12332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175058.4 NP_778228.3 23 4019
XM_017017241.1 XP_016872730.1 25 4230
XM_017017242.1 XP_016872731.1 24 4110
XM_017017243.1 XP_016872732.1 23 4029
GenBank mRNA BC033239.1 - 14 2899
BC041452.1 - 5 1357
BC071599.1 - 23 4015 30343461
BX648885.1 - 2 2802

View Full Product Details