Gene Transcripts
- Gene Symbol
- SCRN1
- Entrez Gene ID
- 9805
- Gene Name
- secernin 1
- Gene Aliases
- SES1
- Chromosome Location
- Chr. 7: 29920103 - 29990289 on Build GRCh38
- UniGene ID
- Hs.520740
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145513.1 |
NP_001138985.1
|
2, 3 |
338 |
|
NM_001145514.1 |
NP_001138986.1
|
2, 3 |
270 |
|
NM_014766.4 |
NP_055581.3
|
2, 3 |
330 |
|
GenBank mRNA |
AB071705.1 |
- |
2, 3 |
220 |
|
AK289514.1 |
- |
1, 2 |
365 |
|
AK293462.1 |
- |
2, 3 |
282 |
|
AK295172.1 |
- |
2, 3 |
270 |
|
BC040492.1 |
- |
2, 3 |
282 |
5314711 |
D83777.2 |
- |
2, 3 |
275 |
|
DC307846.1 |
- |
2, 3 |
339 |
|
DC370097.1 |
- |
2, 3 |
330 |
|
View Full Product Details