Gene Transcripts

Gene Symbol
Adam6b
Entrez Gene ID
238405
Gene Name
a disintegrin and metallopeptidase domain 6B
Gene Aliases
4930523C11Rik
Chromosome Location
Chr. 12: 113489565 - 113491835 on Build GRCm38
UniGene ID
Mm.442216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009545.1 NP_001009545.1 1 855
GenBank mRNA AK029742.1 - 1 910
AK076580.1 - 1 910
BC111877.1 - 1 875 40059665

View Full Product Details