Gene Transcripts

Gene Symbol
Kdm2a
Entrez Gene ID
225876
Gene Name
lysine (K)-specific demethylase 2A
Gene Aliases
100043628, 5530401A10Rik, AA589516, AW536790, Cxxc8, Fbl11, Fbl7, Fbxl11, Gm4560, Jhdm1, Jhdm1a, lalina
Chromosome Location
Chr. 19: 4316144 - 4398280 on Build GRCm38
UniGene ID
Mm.31941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001984.2 NP_001001984.2 10 1772
GenBank mRNA AK037157.1 - 11 1669
AK155866.1 - 10 1777
BC037117.1 - 3 321
BC076576.1 - 10 1338 6840591

View Full Product Details