Gene Transcripts

Gene Symbol
Dek
Entrez Gene ID
306817
Gene Name
DEK proto-oncogene
Gene Aliases
Chromosome Location
Chr. 17: 17965858 - 17987836 on Build Rnor_6.0
UniGene ID
Rn.25099
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004255.1 NP_001004255.1 8 915
XM_006253756.2 XP_006253818.1 8 938
GenBank mRNA BC079344.1 - 8 915

View Full Product Details