Gene Transcripts

Gene Symbol
OLFM3
Entrez Gene ID
118427
Gene Name
olfactomedin 3
Gene Aliases
NOE3, NOELIN3, OPTIMEDIN
Chromosome Location
Chr. 1: 101800028 - 101997234 on Build GRCh38
UniGene ID
Hs.484475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288821.1 NP_001275750.1 6 970
NM_001288823.1 NP_001275752.1 7 1282
NM_058170.3 NP_477518.2 6 1221
NR_110210.1 8 2019
NR_110211.1 7 1958
NR_110212.1 7 1707
XM_017000240.1 XP_016855729.1 7 1125
GenBank mRNA AF397392.2 - 6 863
AF397393.2 - 7 1600
AF397394.1 - 6 892
AF397395.1 - 7 1629
AF397396.1 - 7 953
AF397397.1 - 8 1690
AK095551.1 - 4 447
AK095724.1 - 6 1017
AK300870.1 - 6 970
AY358722.1 - 6 934
BC022531.1 - 6 979 4812035

View Full Product Details