Gene Transcripts

Gene Symbol
OR7D2
Entrez Gene ID
162998
Gene Name
olfactory receptor family 7 subfamily D member 2
Gene Aliases
HTPCRH03, OR19-10, OR19-4
Chromosome Location
Chr. 19: 9185594 - 9188817 on Build GRCh38
UniGene ID
Hs.531755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175883.2 NP_787079.1 1 1295
GenBank mRNA AK095468.1 - 1 1295

View Full Product Details