Gene Transcripts

Gene Symbol
Cyp2d12
Entrez Gene ID
380997
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 12
Gene Aliases
9030605E09Rik
Chromosome Location
Chr. 15: 82554960 - 82559413 on Build GRCm38
UniGene ID
Mm.482248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201360.1 NP_958748.1 1 210
GenBank mRNA AK078880.1 - 1 210
AK165474.1 - 219
BC026417.1 - 2 137
BC089596.1 - 1 219
BC139169.1 - 1 194 8862191
BC139170.1 - 1 194 8862192
BC145567.1 - 1 194 9054132

View Full Product Details