Gene Transcripts

Gene Symbol
RRH
Entrez Gene ID
10692
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
Gene Aliases
Chromosome Location
Chr. 4: 109827994 - 109844707 on Build GRCh38
UniGene ID
Hs.658310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006583.2 NP_006574.1 2, 3 339
GenBank mRNA AF012270.1 - 2, 3 355
BC128120.1 - 2, 3 330 40112275
BC128121.1 - 2, 3 330 40112276
BC128401.1 - 2, 3 330 40112297

View Full Product Details