Gene Transcripts

Gene Symbol
Plekha8
Entrez Gene ID
231999
Gene Name
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8
Gene Aliases
6330400G01, AA517676, BC052360, Fapp-2, Fapp2
Chromosome Location
Chr. 6: 54595111 - 54645823 on Build GRCm38
UniGene ID
Mm.274693
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001335.2 NP_001001335.1 5 756
NM_001164361.1 NP_001157833.1 5 851
GenBank mRNA AK078040.1 - 5 852
AK170592.1 - 5 618
BC052360.1 - 5 756 6704789

View Full Product Details