Gene Transcripts

Gene Symbol
Clec4a1
Entrez Gene ID
269799
Gene Name
C-type lectin domain family 4, member a1
Gene Aliases
BC049354, Dcir4
Chromosome Location
Chr. 6: 122921848 - 122934619 on Build GRCm38
UniGene ID
Mm.297275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199311.2 NP_955015.1 3 416
GenBank mRNA AY397675.1 - 3 351
BC049354.1 - 3 406 6310008
CK626014.1 - 3 416

View Full Product Details