Gene Transcripts

Gene Symbol
Tpte
Entrez Gene ID
234129
Gene Name
transmembrane phosphatase with tensin homology
Gene Aliases
Pten2
Chromosome Location
Chr. 8: 22283441 - 22371418 on Build GRCm38
UniGene ID
Mm.450553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199257.2 NP_954866.2 4 528
GenBank mRNA AJ311311.1 - 4 531
AJ311312.1 - 4 616
AJ311313.1 - 4 531
BC107329.2 - 4 323 40054415

View Full Product Details