Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015397.3 | NP_056212.1 | 2 | 457 | |
XM_011517843.1 | XP_011516145.1 | 2 | 284 | ||
GenBank mRNA | AB067479.1 | - | 2 | 377 | |
AK000354.1 | - | 2 | 235 | ||
AK292935.1 | - | 2 | 263 | ||
BC008893.2 | - | 1 | 20 | ||
BC063823.1 | - | 2 | 375 | 6184796 | |
BG718613.1 | - | 2 | 428 | ||
DQ518909.1 | - | 2 | 377 |