Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015490.3 | NP_056305.1 | 3 | 291 | |
GenBank mRNA | AF274863.1 | - | 3 | 287 | |
AK289975.1 | - | 3 | 273 | ||
AK294551.1 | - | 3 | 407 | ||
BC034946.1 | - | 3 | 259 | ||
BC044569.1 | - | 3 | 252 | ||
BC053949.1 | - | 3 | 252 | ||
BC071580.1 | - | 3 | 312 | ||
BC144313.1 | - | 2 | 232 | 9052833 | |
BX640914.1 | - | 3 | 337 |