Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318508.1 | NP_001305437.1 | 2, 3 | 163 | |
NM_015962.4 | NP_057046.1 | 2, 3 | 127 | ||
XM_005267730.3 | XP_005267787.1 | 2, 3 | 129 | ||
GenBank mRNA | AF132969.1 | - | 1, 2 | 76 | |
AK290666.1 | - | 2, 3 | 108 | ||
BX248271.1 | - | 2, 3 | 89 | ||
BX489015.1 | - | 2, 3 | 129 | ||
BX647810.1 | - | 2, 3 | 153 |