Gene Transcripts

Gene Symbol
HDDC2
Entrez Gene ID
51020
Gene Name
HD domain containing 2
Gene Aliases
C6orf74, CGI-130, NS5ATP2, dJ167O5.2
Chromosome Location
Chr. 6: 125274934 - 125302141 on Build GRCh38
UniGene ID
Hs.32826
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016063.2 NP_057147.2 6 1156
GenBank mRNA AF151888.1 - 5 913

View Full Product Details