Gene Transcripts
- Gene Symbol
- VPS28
- Entrez Gene ID
- 51160
- Gene Name
- VPS28, ESCRT-I subunit
- Gene Aliases
- Chromosome Location
- Chr. 8: 144423601 - 144428563 on Build GRCh38
- UniGene ID
- Hs.418175
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016208.3 |
NP_057292.1
|
4, 5 |
215 |
|
NM_183057.2 |
NP_898880.1
|
4, 5 |
215 |
|
XM_005272323.3 |
XP_005272380.1
|
4, 5 |
207 |
|
XM_005272324.3 |
XP_005272381.1
|
4, 5 |
207 |
|
GenBank mRNA |
AF182844.1 |
- |
3, 4 |
167 |
|
AF316887.1 |
- |
3, 4 |
149 |
|
AK026865.1 |
- |
4, 5 |
165 |
|
BC006485.1 |
- |
3, 4 |
151 |
3535787 |
BC019321.1 |
- |
3, 4 |
151 |
2822758 |
BC050713.2 |
- |
4, 5 |
163 |
5741129 |
BU658837.1 |
- |
4, 5 |
508 |
|
KU178635.1 |
- |
3, 4 |
106 |
|
KU178636.1 |
- |
3, 4 |
106 |
|
R72904.1 |
- |
3, 4 |
97 |
|
View Full Product Details