Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001606.4 | NP_001597.2 | 5 | 475 | |
NM_212533.2 | NP_997698.1 | 5 | 467 | ||
XM_006716996.3 | XP_006717059.1 | 4 | 409 | ||
GenBank mRNA | AF178941.1 | - | 4 | 377 | |
AF327657.1 | - | 4 | 333 | ||
AK308738.1 | - | 5 | 372 | ||
BC029282.1 | - | 4 | 433 | ||
BC064542.1 | - | 5 | 363 | ||
BC144588.1 | - | 4 | 457 | ||
BM921499.1 | - | 4 | 423 | ||
CD676110.1 | - | 5 | 413 |