Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001552.4 | NP_001001552.3 | 4 | 547 | |
NM_001199050.1 | NP_001185979.1 | 6 | 689 | ||
NM_001199051.1 | NP_001185980.1 | 5 | 566 | ||
NM_001199052.1 | NP_001185981.1 | 3 | 424 | ||
NR_037583.1 | 4 | 501 | |||
XM_011510160.2 | XP_011508462.1 | 7 | 777 | ||
XM_011510161.2 | XP_011508463.1 | 7 | 765 | ||
XM_011510162.2 | XP_011508464.1 | 7 | 757 | ||
XM_011510163.2 | XP_011508465.1 | 6 | 657 | ||
XM_011510164.2 | XP_011508466.1 | 8 | 844 | ||
XM_011510165.2 | XP_011508467.1 | 8 | 816 | ||
GenBank mRNA | AB084764.1 | - | 3 | 391 | |
AB084765.1 | - | 4 | 468 | ||
AB085610.1 | - | 4 | 514 | ||
AB096677.1 | - | 6 | 656 | ||
AB096678.1 | - | 4 | 468 | ||
AB096679.1 | - | 5 | 533 | ||
AB096680.1 | - | 5 | 591 | ||
AB096681.1 | - | 4 | 514 | ||
AB096682.1 | - | 3 | 391 | ||
AK026628.1 | - | 4 | 514 | ||
AK302392.1 | - | 6 | 691 | ||
BC036636.2 | - | 6 | 659 | 5296437 |