Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005937.3 | NP_005928.2 | 11, 12 | 1841 | |
XM_006721911.2 | XP_006721974.1 | 11, 12 | 1763 | ||
XM_017024648.1 | XP_016880137.1 | 6, 7 | 1366 | ||
XM_017024649.1 | XP_016880138.1 | 11, 12 | 1841 | ||
GenBank mRNA | AB209633.1 | - | 11, 12 | 1816 | |
BC007237.1 | - | 3, 4 | 792 | ||
BC007859.2 | - | 1, 2 | 192 | ||
U07932.1 | - | 11, 12 | 1828 |