Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr. 13: 34942287 - 35672737 on Build GRCh38
UniGene ID
Hs.491172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204197.1 NP_001191126.1 9 991
NM_015678.4 NP_056493.3 49 8021
XM_005266346.4 XP_005266403.1 49 8021
XM_005266347.4 XP_005266404.2 49 8021
XM_005266348.4 XP_005266405.2 48 8012
XM_006719803.3 XP_006719866.1 49 8021
XM_006719805.3 XP_006719868.1 48 8012
XM_006719806.3 XP_006719869.1 48 8012
XM_011535046.2 XP_011533348.1 41 6152
XM_017020544.1 XP_016876033.1 49 8003
XM_017020545.1 XP_016876034.1 49 8003
XM_017020546.1 XP_016876035.1 49 7309
GenBank mRNA AB046764.1 - 16 1733
AF072371.1 - 3 208
AF467288.1 - 50 7693
AK091469.1 - 20 2180
AK122888.1 - 19 2534
AK127791.1 - 26 3372
AK294737.1 - 9 991
AK297773.1 - 10 1089
AK315939.1 - 9 988
AL137748.1 - 3 633
BX640760.1 - 25 3351

View Full Product Details