Gene Transcripts

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
Chromosome Location
Chr. 4: 168216291 - 168318807 on Build GRCh38
UniGene ID
Hs.591710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017631.5 NP_060101.3 36 5134
XM_011532103.2 XP_011530405.1 36 5134
GenBank mRNA AK000042.1 - 8 935
AK026333.1 - 4 400
AK223184.1 - 8 935
AK225191.1 - 8 922
AL137464.1 - 3 320
BC020601.1 - 4 350
BC038115.2 - 36 5079 5261001
BC074781.2 - 8 913
BX648655.1 - 16 1976

View Full Product Details