Gene Transcripts

Gene Symbol
TP53AIP1
Entrez Gene ID
63970
Gene Name
tumor protein p53 regulated apoptosis inducing protein 1
Gene Aliases
P53AIP1
Chromosome Location
Chr. 11: 128934732 - 128944233 on Build GRCh38
UniGene ID
Hs.160953
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195194.1 NP_001182123.1 3 550
NM_001251964.1 NP_001238893.1 2 3081
NM_022112.2 NP_071395.2 4 1228
XM_017018113.1 XP_016873602.1 4 1993
XM_017018114.1 XP_016873603.1 4 1987
XM_017018116.1 XP_016873605.1 4 5258
GenBank mRNA AB045830.1 - 4 756
AB045832.1 - 2 2609

View Full Product Details