Gene Transcripts
- Gene Symbol
- GFM1
- Entrez Gene ID
- 85476
- Gene Name
- G elongation factor mitochondrial 1
- Gene Aliases
- COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1
- Chromosome Location
- Chr. 3: 158644497 - 158692572 on Build GRCh38
- UniGene ID
- Hs.518355
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308164.1 |
NP_001295093.1
|
8, 9 |
1168 |
|
NM_001308166.1 |
NP_001295095.1
|
7, 8 |
1111 |
|
NM_024996.5 |
NP_079272.4
|
7, 8 |
1111 |
|
XM_006713795.1 |
XP_006713858.1
|
6, 7 |
1025 |
|
GenBank mRNA |
AF309777.1 |
- |
7, 8 |
1111 |
|
AF367998.1 |
- |
7, 8 |
1111 |
|
AK000780.1 |
- |
7, 8 |
1111 |
|
AK022724.1 |
- |
2, 3 |
213 |
|
AK092293.1 |
- |
6, 7 |
717 |
|
AK315031.1 |
- |
7, 8 |
1050 |
|
BC049210.1 |
- |
7, 8 |
1075 |
5574223 |
KU178816.1 |
- |
7, 8 |
1004 |
|
KU178817.1 |
- |
6, 7 |
883 |
|
View Full Product Details