Gene Transcripts

Gene Symbol
PLK4
Entrez Gene ID
10733
Gene Name
polo like kinase 4
Gene Aliases
MCCRP2, SAK, STK18
Chromosome Location
Chr. 4: 127880861 - 127899224 on Build GRCh38
UniGene ID
Hs.172052
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190799.1 NP_001177728.1 4 1525
NM_001190801.1 NP_001177730.1 5 1444
NM_014264.4 NP_055079.3 5 1621
XM_017007662.1 XP_016863151.1 5 1780
GenBank mRNA AB006972.1 - 5 1535
AK302858.1 - 5 1444
AK303399.1 - 4 1459
AK314238.1 - 5 1589
BC036023.1 - 5 1589 5273226
Y13115.1 - 5 1487

View Full Product Details