Gene Transcripts

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
Chromosome Location
Chr. 12: 110372900 - 110403730 on Build GRCh38
UniGene ID
Hs.719935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137664.1 NP_001131136.1 8, 9 1243
NM_016238.2 NP_057322.2 8, 9 1243
XM_005253903.3 XP_005253960.1 7, 8 1109
XM_011538454.2 XP_011536756.1 8, 9 1471
XM_017019426.1 XP_016874915.1 8, 9 1238
XM_017019427.1 XP_016874916.1 8, 9 2037
XM_017019428.1 XP_016874917.1 7, 8 1126
GenBank mRNA AF191340.1 - 8, 9 1140
AK021809.1 - 8, 9 1222
AL137586.1 - 2, 3 297
BC002941.2 - 3, 4 387
BC009498.2 - 6, 7 813
BC017316.2 - 4, 5 589
BC040130.1 - 8, 9 1158
BC098264.1 - 8, 9 1139
BC098295.1 - 8, 9 1139
BC098362.1 - 8, 9 1139
BC099732.1 - 8, 9 1139
BC111798.1 - 8, 9 1139
BC141848.1 - 8, 9 1193 8860346
BC148236.2 - 8, 9 1140 40024981

View Full Product Details