Gene Transcripts

Gene Symbol
TPRA1
Entrez Gene ID
131601
Gene Name
transmembrane protein adipocyte associated 1
Gene Aliases
GPR175, TMEM227, TPRA40
Chromosome Location
Chr. 3: 127573064 - 127598234 on Build GRCh38
UniGene ID
Hs.6418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136053.2 NP_001129525.1 7 918
NM_001142646.2 NP_001136118.1 7 918
NR_073377.1 7 918
XM_006713495.3 XP_006713558.1 8 1099
XM_006713496.2 XP_006713559.1 7 776
XM_006713497.3 XP_006713560.1 8 1143
XM_011512422.1 XP_011510724.1 7 692
XM_017005707.1 XP_016861196.1 8 882
XM_017005708.1 XP_016861197.1 8 912
XM_017005709.1 XP_016861198.1 8 1013
GenBank mRNA AB037108.1 - 6 842
AF289568.1 - 7 865
BC017058.1 - 7 687 3904719
BC050711.2 - 7 812 6168632
CD580484.2 - 7 653

View Full Product Details