Gene Transcripts

Gene Symbol
RIMBP2
Entrez Gene ID
23504
Gene Name
RIMS binding protein 2
Gene Aliases
PPP1R133, RBP2, RIM-BP2
Chromosome Location
Chr. 12: 130396133 - 130716361 on Build GRCh38
UniGene ID
Hs.657441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015347.4 NP_056162.4 19 3602
XM_017019110.1 XP_016874599.1 22 3976
GenBank mRNA AB002316.1 - 19 3602

View Full Product Details