Gene Transcripts

Gene Symbol
C2cd2l
Entrez Gene ID
300666
Gene Name
C2CD2-like
Gene Aliases
Tmem24
Chromosome Location
Chr. 8: 48641792 - 48652115 on Build Rnor_6.0
UniGene ID
Rn.57410
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011996.1 NP_001011996.1 2 797
XM_006242917.2 XP_006242979.1 2 848
XM_006242918.2 XP_006242980.1 2 848
XM_006242919.2 XP_006242981.1 2 848
XM_006242920.2 XP_006242982.1 2 848
XM_008766156.1 XP_008764378.1 2 848
XM_008766157.1 XP_008764379.1 2 400
GenBank mRNA BC085925.1 - 2 797

View Full Product Details