Gene Transcripts

Gene Symbol
Ppapdc3
Entrez Gene ID
296635
Gene Name
phosphatidic acid phosphatase type 2 domain containing 3
Gene Aliases
RGD1305821
Chromosome Location
Chr. 3: 11114551 - 11130427 on Build Rnor_6.0
UniGene ID
Rn.24087
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012349.2 NP_001012349.1 2 1427
GenBank mRNA BC089948.1 - 2 1427

View Full Product Details