Gene Transcripts

Gene Symbol
Tm7sf2
Entrez Gene ID
293688
Gene Name
transmembrane 7 superfamily member 2
Gene Aliases
Chromosome Location
Chr. 1: 221426017 - 221430340 on Build Rnor_6.0
UniGene ID
Rn.8313
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013071.1 NP_001013089.1 7, 8 987
GenBank mRNA BC091237.1 - 7, 8 987

View Full Product Details