Gene Transcripts

Gene Symbol
Pgrmc1
Entrez Gene ID
291948
Gene Name
progesterone receptor membrane component 1
Gene Aliases
25-Dx, 25Dx, MPR, VEMA
Chromosome Location
Chr. X: 123205869 - 123213880 on Build Rnor_6.0
UniGene ID
Rn.102356
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021766.1 NP_068534.1 3 984
GenBank mRNA AF163321.1 - 3 941
BC062073.1 - 3 944 5599449
U63315.1 - 3 984

View Full Product Details