Gene Transcripts

Gene Symbol
Olr6
Entrez Gene ID
365175
Gene Name
olfactory receptor 6
Gene Aliases
Chromosome Location
Chr. 1: 70480941 - 70481879 on Build Rnor_6.0
UniGene ID
Rn.142003
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000539.1 NP_001000539.1 1 264

View Full Product Details