Gene Transcripts
- Gene Symbol
- OSBPL10
- Entrez Gene ID
- 114884
- Gene Name
- oxysterol binding protein like 10
- Gene Aliases
- ORP10, OSBP9
- Chromosome Location
- Chr. 3: 31660825 - 31981850 on Build GRCh38
- UniGene ID
- Hs.150122
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001174060.1 |
NP_001167531.1
|
1, 2 |
960 |
|
NM_017784.4 |
NP_060254.2
|
1, 2 |
960 |
|
XM_005264843.3 |
XP_005264900.1
|
1, 2 |
736 |
|
XM_005264844.4 |
XP_005264901.1
|
1, 2 |
736 |
|
XM_017005668.1 |
XP_016861157.1
|
1, 2 |
603 |
|
GenBank mRNA |
AF346291.1 |
- |
1, 2 |
406 |
|
AF392451.1 |
- |
2, 3 |
670 |
|
AK304067.1 |
- |
1, 2 |
413 |
|
AL833247.1 |
- |
1, 2 |
63 |
|
BC003168.1 |
- |
1, 2 |
364 |
2905981 |
View Full Product Details