Gene Transcripts

Gene Symbol
Lrrc41
Entrez Gene ID
362566
Gene Name
leucine rich repeat containing 41
Gene Aliases
RGD1311221
Chromosome Location
Chr. 5: 134927609 - 134948530 on Build Rnor_6.0
UniGene ID
Rn.1938
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009710.1 NP_001009710.1 4 777
XM_006238682.2 XP_006238744.1 4 1162
GenBank mRNA BC087037.1 - 4 777

View Full Product Details