Gene Transcripts

Gene Symbol
LDLRAD1
Entrez Gene ID
388633
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
Chromosome Location
Chr. 1: 54007298 - 54018186 on Build GRCh38
UniGene ID
Hs.439583
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010978.3 NP_001010978.2 6 676
NM_001276392.1 NP_001263321.1 4 495
NM_001276393.1 NP_001263322.1 5 547
NM_001276394.1 NP_001263323.1 4 409
NM_001276395.1 NP_001263324.1 5 516
XM_011541446.1 XP_011539748.1 6 656
GenBank mRNA BC103740.1 - 4 397
BC127708.1 - 6 673 40127476
BC127709.1 - 4 492 40127477
BC140760.1 - 5 491 9021628
BC144466.1 - 5 491 9052992
BC171743.1 - 5 460

View Full Product Details