Gene Transcripts

Gene Symbol
Tssc1
Entrez Gene ID
362721
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
Chromosome Location
Chr. 6: 48021771 - 48135241 on Build Rnor_6.0
UniGene ID
Rn.119114
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012192.1 NP_001012192.1 9 1379
GenBank mRNA BC087633.1 - 9 1379

View Full Product Details