Gene Transcripts

Gene Symbol
BAK1
Entrez Gene ID
578
Gene Name
BCL2 antagonist/killer 1
Gene Aliases
BAK, BAK-LIKE, BCL2L7, CDN1
Chromosome Location
Chr. 6: 33572546 - 33580293 on Build GRCh38
UniGene ID
Hs.485139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001188.3 NP_001179.1 3 395
XM_011514779.2 XP_011513081.1 4 506
GenBank mRNA AF520590.1 - 2 95
AK292093.1 - 4 433
AK293547.1 - 3 325
AK303422.1 - 3 374
BC004431.1 - 1 306 2819507
BC110337.1 - 3 305 5755200
CB142221.1 - 3 395
CR457419.1 - 2 95
CR627020.1 - 1 4169
KU177934.1 - 2 95
KU177935.1 - 2 95
U16811.1 - 3 295
U23765.1 - 3 159
X84213.1 - 3 287

View Full Product Details