Gene Transcripts

Gene Symbol
Plet1
Entrez Gene ID
363060
Gene Name
placenta expressed transcript 1
Gene Aliases
Chromosome Location
Chr. 8: 54925666 - 54937007 on Build Rnor_6.0
UniGene ID
Rn.39346
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014209.1 NP_001014231.1 4 526
GenBank mRNA BC088858.1 - 4 526
FQ228964.1 - 4 549

View Full Product Details