Gene Transcripts

Gene Symbol
IL17C
Entrez Gene ID
27189
Gene Name
interleukin 17C
Gene Aliases
CX2, IL-17C
Chromosome Location
Chr. 16: 88638593 - 88640474 on Build GRCh38
UniGene ID
Hs.278911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013278.3 NP_037410.1 3 1012
GenBank mRNA AF152099.1 - 3 1011
AY358471.1 - 3 1011

View Full Product Details