Gene Transcripts
- Gene Symbol
- TTC7A
- Entrez Gene ID
- 57217
- Gene Name
- tetratricopeptide repeat domain 7A
- Gene Aliases
- GIDID, MINAT, TTC7
- Chromosome Location
- Chr. 2: 46915931 - 47076136 on Build GRCh38
- UniGene ID
- Hs.370603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288951.1 |
NP_001275880.1
|
6, 7 |
1209 |
|
NM_001288953.1 |
NP_001275882.1
|
7, 8 |
1200 |
|
NM_020458.3 |
NP_065191.2
|
6, 7 |
1209 |
|
XM_005264439.3 |
XP_005264496.1
|
7, 8 |
1362 |
|
XM_011532998.2 |
XP_011531300.1
|
8, 9 |
1427 |
|
XM_011532999.2 |
XP_011531301.1
|
6, 7 |
1152 |
|
XM_011533000.2 |
XP_011531302.1
|
3, 4 |
284 |
|
XM_017004524.1 |
XP_016860013.1
|
6, 7 |
1153 |
|
XM_017004525.1 |
XP_016860014.1
|
6, 7 |
694 |
|
XM_017004526.1 |
XP_016860015.1
|
6, 7 |
1153 |
|
XM_017004527.1 |
XP_016860016.1
|
6, 7 |
1304 |
|
XM_017004528.1 |
XP_016860017.1
|
7, 8 |
1369 |
|
XM_017004529.1 |
XP_016860018.1
|
6, 7 |
1152 |
|
XM_017004530.1 |
XP_016860019.1
|
6, 7 |
1153 |
|
GenBank mRNA |
AB032966.2 |
- |
7, 8 |
789 |
|
AK056464.1 |
- |
7, 8 |
1172 |
|
AL834383.1 |
- |
6, 7 |
1209 |
|
BC065554.1 |
- |
3, 4 |
327 |
|
BC111487.1 |
- |
6, 7 |
1097 |
6041916 |
View Full Product Details