Gene Transcripts

Gene Symbol
Hyi
Entrez Gene ID
68180
Gene Name
hydroxypyruvate isomerase homolog (E. coli)
Gene Aliases
2700033B16Rik, 6430559E15Rik, HT036
Chromosome Location
Chr. 4: 118359999 - 118362744 on Build GRCm38
UniGene ID
Mm.193212
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026601.3 NP_080877.3 7 858
GenBank mRNA AK004328.1 - 6 446
AK020099.1 - 7 732
BC024619.1 - 7 846 4923664
BC100401.1 - 7 822
BX525112.1 - 5 299
CB952524.1 - 5 363

View Full Product Details