Gene Transcripts

Gene Symbol
Zdhhc18
Entrez Gene ID
503610
Gene Name
zinc finger, DHHC domain containing 18
Gene Aliases
Chromosome Location
Chr. 4: 133605301 - 133633480 on Build GRCm38
UniGene ID
Mm.331948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017968.2 NP_001017968.2 3 611
GenBank mRNA AY668950.1 - 2 190
BC145714.1 - 2 242
BC145716.1 - 2 242

View Full Product Details