Gene Transcripts

Gene Symbol
Lrrc17
Entrez Gene ID
502715
Gene Name
leucine rich repeat containing 17
Gene Aliases
Chromosome Location
Chr. 4: 10108184 - 10138652 on Build Rnor_6.0
UniGene ID
Rn.207235
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025155.2 NP_001020326.1 4 1944
XM_008762657.1 XP_008760879.1 4 2232
GenBank mRNA BC079098.1 - 4 1821

View Full Product Details